Shipper, EAS Ambient, Small Transport Box, Therapak®
Catalog #: 10029-410
Supplier: Therapak, LLC
Specifications
- Description:Small transport box, specimen bag, absorbent
- Cat. no.:10029-410
- Supplier No.:56880
Specifications
About this item
The small transport box is designed for shipping one standard or larger specimen container (30 mL to 60 mL) of liquid specimen.
Shipped with pre-assembled boxes, these transport shippers also include: small transport box (ID: 5.25 x 4 x 3.75"; OD: 6 x 4.25 x 4"), specimen transport bag, absorbent material, Exempt Animal Specimen label, biohazard symbol label and cushioning material where applicable.
Customers Who Bought This Also Bought