Shipper, EHS Combo, Therapak®
Catalog #: 10029-406
Supplier: Therapak, LLC
Specifications
- Description:EHS combo shipping system
- Cat. no.:10029-406
- Supplier No.:56878
Specifications
About this item
EHS Combination specimen shipping system features Therapak®'s standard small cooler (for frozen specimen section) nested underneath an insulated envelope (for ambient specimen section).
Frozen specimens are packed inside a 6x9" zip-style biohazard bag, including an Aqui-Pak™ segmented absorbent pouch that holds up to four tubes, and placed in the small insulated cooler (ID: 8 x 6 x 8”) with the appropriate amount of dry ice.
Ambient section includes 6 x 9" biohazard bag and Aqui-Pak™ segmented absorbent pouch plus an ambient gel wrap to maintain the temperature environment.
System includes: cushioning material, Exempt Human Specimen label, biohazard label, and dry ice label. Add dry ice and sample tubes to ship.
Customers Who Bought This Also Bought