Shipper, EHS Ambient for 6 Tubes, Therapak®
Catalog #: 10029-398
Supplier: Therapak, LLC
Specifications
- Description:Medium transport box, specimen bag, up to 6 tubes
- Cat. no.:10029-398
- Supplier No.:56874
Specifications
About this item
Ambient shipping system features: medium transport box (ID: 7 x 4 x 4"; OD: 8 x 4.25 x 4.25"), specimen transport bag, Aqui-Pak segmented absorbent pouch able to hold up to six standard blood or urine tubes, Exempt Human Specimen label and biohazard symbol label.
Customers Who Bought This Also Bought