Shipper, EHS Ambient for 4 Tubes, Therapak®
Catalog #: 10029-396
Supplier: Therapak, LLC
Specifications
- Description:Small transport box, specimen bag, up to 4 tubes
- Cat. no.:10029-396
- Supplier No.:56873
Specifications
About this item
Ambient shipping system features: small transport box (ID: 5.25 x 4 x 3.75"; OD: 6 x 4.25 x 4"), specimen transport bag, Aqui-Pak™ segmented absorbent pouch to hold up to four standard blood or urine tubes, Exempt Human Specimen label, biohazard symbol label and cushioning material where applicable.
Customers Who Bought This Also Bought