Shipper, Category B Ambient, 95 kPa Bag, Tube Shuttle, Therapak®
Catalog #: 10029-266
Supplier: Therapak, LLC
Specifications
- Description:Cat B ambient 95 kpa bag, tube shuttle
- Cat. no.:10029-266
- Interior Dimensions:7×4×4"
- Exterior Dimensions:8×4.25×4.25"
- Supplier No.:37802
Specifications
About this item
The ambient transport system for blood and urine tube transport includes 95kPa bag, TearTech tube shuttle, absorbent sheet, cushioning material, list of contents card, biohazard symbol label, Biological Substance Category B label, and medium transport box.
Holds up to six tubes.
Customers Who Bought This Also Bought