Shipper, Category B Combo, Frozen/Refrigerated, Therapak®
Catalog #: 10029-276
Supplier: Therapak, LLC
Specifications
- Description:Cat B combo, frozen/refrigerated
- Cat. no.:10029-276
- Supplier No.:37810
Specifications
About this item
Combination Side by Side shipping system utilizes two small coolers situated in one outer box
Inner packaging consists of a medium canister with an Aqui-Pak™ 6-tube absorbent pouch (for Frozen section). The refrigerated section consists of a medium canister with an Aqui-Pak™ 6-tube absorbent pouch and four cold packs. Other materials include orientation labels, list of contents card, biohazard symbol label, frozen specimen label, dry ice label and Biological Substance Category B label. Holds up to six tubes in each section for overnight shipments.
Customers Who Bought This Also Bought