Shipper, Category B Ambient, 95 kPa Medium Canister, Therapak®
Catalog #: 10029-258
Supplier: Therapak, LLC
Specifications
- Description:Cat B ambient, 95kpa Medium canister
- Cat. no.:10029-258
- Interior Dimensions:7×4×4"
- Exterior Dimensions:8×4.25×4.25"
- Supplier No.:37804
Specifications
About this item
The ambient transport system for blood and urine tube transport includes medium 95kPa canister, Aqui-Pak™ segmented absorbent pouch, cushioning material, list of contents card, biohazard symbol label, Biological Substance Category B label, and medium transport box.
Holds up to six tubes.
Customers Who Bought This Also Bought