Samco™ Autoanalyzer Sample Cups, Thermo Scientific
Supplier: Samco
Certificates
About this item
A range of Samco™ autoanalyzer cups suitable for use in a wide range of analyzers.
- High clarity polystyrene
- Precision molded graduations for accurate volume handling
- Resistant to most aqueous solutions
Certifications: FDA Class 1 Exempt Medical Device.
Ordering information: Ambient storage and shipping recommended for containers.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.