Nutra-Gel Diet™, Purified Formula, Rodent, Dry Mix Kit, Bio-Serv
Catalog #: CA89408-236
Supplier: Bio-Serv
Specifications
- Description:Nutra-Gel Dry Mix Kit, Bacon Flavor
- Cat. no.:CA89408-236
- Supplier No.:F5769-KIT
Specifications
About this item
Nutritionally complete, purified, bacon-flavored gel diet that provides both food and water for rodents, and is especially useful in cases involving chewing and mobility impairments, post-op care, and weanlings
- Kit provides the powdered diet and trays to make 4 trays of 16 cubes/tray at 40 g/cube of Nutra-Gel
- Easy to prepare; just add hot water and mix according to directions
- Prepare desired quantities of Nutra-Gel as needed
- Place Nutra-Gel directly on the bedding or in the food hopper
- Nutra-Gel remains moist in the cage 24 to 48 hours, depending on the type of cage system
- Allows incorporation of drugs into the diet for ease of delivery
- Powder is stable for long transit time in delivery
- Nutritionally assayed
- Shelf Life: 6 months, refrigerated
Ingredients: Sucrose, Casein, Corn Starch, Soybean Oil, Gelatin, Cellulose, Mineral Mix, Vitamin Mix, Bacon Flavor, L-Cystine, Xanthan Gum, Sorbic Acid, Choline Bitartrate, tBHQ.
Caloric value: 3.90 kcal/g
Nutritional ash content: 2.2%
Nutritional carbohydrate content: 55.6%
Nutritional fat content: 8.3%
Nutritional fibre content: 4.8%
Nutritional moisture content: <10%
Nutritional protein content: 23.7%
Ordering information: Most mice consume approximately 10-12 g of Nutra-Gel per day. Rats consume approximately 26-60 g of Nutra-Gel per day, depending on strain and age.
Customers Who Bought This Also Bought