Nutra-Gel Diet™, Grain-Based Formula, Rodent, Dry Mix Kit, Bio-Serv
Catalog #: CA89408-234
Supplier: Bio-Serv
Specifications
- Description:Nutra-Gel Dry Mix Kit, Cherry Flavor
- Cat. no.:CA89408-234
Specifications
About this item
Nutritionally complete, grain-based, cherry-flavored gel diet that provides both food and water for rodents, and is especially useful in cases involving chewing and mobility impairments, post-op care, and weanlings
- Kit provides the powdered diet and trays to make 4 trays of 16 cubes/tray at 40 g/cube of Nutra-Gel
- Easy to prepare; just add hot water and mix according to directions
- Prepare desired quantities of Nutra-Gel as needed
- Place Nutra-Gel directly on the bedding or in the food hopper
- Nutra-Gel remains moist in the cage 24 to 48 hours, depending on the type of cage system
- Allows incorporation of drugs into the diet for ease of delivery
- Powder is stable for long transit time in delivery
- Nutritionally assayed
- Shelf Life: 6 months, refrigerated
Ingredients: Ground Wheat, Ground Corn, Dehulled Soybean Meal, Wheat Germ, Fish Meal, Brewer’s Dried Yeast, Corn Gluten Meal, Porcine Animal Fat (preserved with BHA), Soybean Oil, Gelatin, Mineral Mix, Vitamin Mix, Cherry Flavor, DL-Methionine, Choline Chloride, Sorbic Acid.
Caloric value: 3.74 kcal/g
Nutritional ash content: 4.7%
Nutritional carbohydrate content: 48.9%
Nutritional fat content: 8.5%
Nutritional fibre content: 2%
Nutritional moisture content: <10%
Nutritional protein content: 25.4%
Ordering information: Most mice consume approximately 10-12 g of Nutra-Gel per day. Rats consume approximately 26-60 g of Nutra-Gel per day, depending on strain and age.
Customers Who Bought This Also Bought