ReliaPrep™ Non sterile Nylon Syringe Filters, Ahlstrom
Supplier: Ahlstrom
Certificates
About this item
Ahlstrom offers syringe filters for convenient filtration of small sample sizes through membranes.
Nylon (polyamide) membrane is useful in both aqueous and organic solvent fitlration applications.
Customers Who Bought This Also Bought