Softies™ Primate Diet, Certified, Bio-Serv
Catalog #: CA89067-392
Supplier: Bio-Serv
Specifications
- Description:Softies Pellets, Banana Flavor
- Cat. no.:CA89067-392
- Packaging:10 kg/Box
- Supplier No.:F3478
Specifications
About this item
Soft, pelleted, nutritionally complete diet is ideal for chewing difficulties, post-op care or finicky eaters
- Excellent transition diet for weanlings
- Nutritionally assayed
- Can be used with other species
- Shelf Life: 6 months refrigerated
Ingredients: Sucrose, Casein, Whole Milk Powder, Soy Flour, Corn Syrup Solids, Cellulose, Corn Oil, Molasses, Mineral Mix, Wheat Bran, Magnesium Stearate, Acacia Gum, Glycerin, Vitamin Mix, Natural and Artificial Flavors, Choline Chloride, DL-Methionine, Ascorbic Acid, Vitamin E Acetate, BHA, Food Dye.
Caloric value: 3.63 kcal/g
Nutritional ash content: 5.3%
Nutritional carbohydrate content: 44.6%
Nutritional fat content: 10.4%
Nutritional fibre content: 8.5%
Nutritional moisture content: <10%
Nutritional protein content: 22.9%
Vitamin C: 2.7 mg/g
Vitamin D3: 3.6 IU/g
Customers Who Bought This Also Bought