Primilac™ Liquid Infant Primate Diet, Bio-Sev
Catalog #: CA89067-386
Supplier: Bio-Serv
Specifications
- Description:Primilac
- Cat. no.:CA89067-386
- Packaging:1 kg/Bag
- Supplier No.:F0554SP
Specifications
About this item
A milk substitute for infant non-human primates containing selenium, chromium and fluoride, which are not added to human formulas.
- Contains appropriate amounts of protein, fat and carbohydrates for most primates
- Contains a higher level of Vitamin C per serving compared to human formulas
- Easy to prepare; no blender needed. Add water, shake and pour. Use with Bio-Serv's 160 oz. Shake and Pour Bottle or your own mixing vessel.
- Nutritionally assayed
Shelf Life: 6 months refrigerated. If prepared, refrigerate unused portion up to 3 days. Prepared diet should not be kept more than 48 hours at room temperature.
Ingredients: Lactose, Casein, Maltodextrin, Coconut Oil, Corn Oil, Chicken Fat, Mineral Mix, Cellulose, Lecithin, Suspending Aids, Vitamin Mix, Choline Dihydrogen Citrate, Banana Flavor, L-Cystine, BHT.
Nutritional carbohydrate content: 54.5 kcals/6 oz.
Nutritional fat content: 67.8 kcals/6 oz.
Nutritional protein content: 22.5 kcals/6 oz.
Total caloric profile: 144.7 kcals/6 oz.
Customers Who Bought This Also Bought