Supreme Mini-Treats™, Certified, Bio-Serv
Supplier: Bio-Serv
Certificates
About this item
Nutritionally complete 1 g treats that compliment enrichment programs. Excellent for training and conditioning or as a reward following a less desirable procedure.
- Appetizing aroma helps to deter rodent maternal cannibalism
- chocolate and very berry flavor recommended.
- Excellent source of vitamin C
- Nutritionally assayed
- Shelf Life: 12 months, cool dry storage
Ingredients: Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corn Oil, Corn Syrup, Mineral Mix, Choline Bitartrate, Natural and Artificial Flavors, Vitamin Mix, Calcium Silicate, Magnesium Stearate, L-Cystine, Ascorbic Acid, Food Dye (Very Berry Flavor Only).
Caloric value: 3.46 kcal/Pellet
Nutritional ash content: 5.2%
Nutritional carbohydrate content: 53.0%
Nutritional fat content: 6.1%
Nutritional fibre content: 11.2%
Nutritional moisture content:<10%
Nutritional protein content: 19.7%
Vitamin C:1.1 mg/Pellet
Vitamin D3: 2.0 mg/Pellet
Certifications: Certified (Contaminant Screened).
Customers Who Bought This Also Bought