Ultra-2 Non-Polar GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
Certificates
About this item
HP-Ultra-2 phase
Ordering information: These columns are suitable replacements for HP-Ultra-2.
Specifications
- Catalog No:
- 10498-866
- 10498-868
- 10498-870
- 10498-872
- 10498-858
- 10498-860
- 10498-862
- 10498-864
- 10498-856
- Description:
- 10498-866
- 10498-868
- 10498-870
- 10498-872
- 10498-858
- 10498-860
- 10498-862
- 10498-864
- 10498-856
- Temperature limit:
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C