GsBP®-225MS Mid-Polar GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
Certificates
About this item
Equivalent to: DB-225, DB-225MS, Rtx-225.
- 25% cyanopropyl, 25% phenyl, 50% methyl polysiloxane phase
- Mid-polar
- Close USP G7
Specifications
- Catalog No:
- 10499-964
- 10499-966
- 10499-968
- 10499-970
- 10499-980
- 10499-974
- 10499-976
- 10499-978
- Description:
- 10499-964
- 10499-966
- 10499-968
- 10499-970
- 10499-980
- 10499-974
- 10499-976
- 10499-978
- Temperature limit:
- 40 to 220/240°C
- 40 to 220/240°C
- 40 to 220/240°C
- 40 to 220/240°C
- 40 to 220°C
- 40 to 220/240°C
- 40 to 220°C
- 40 to 220°C