GsBP®-1701 Mid-Polar GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
Certificates
About this item
Typical Application: 5% Fragrance Materials Association Mix, Acrylate Impurities II, Antidepressants (Basic Drugs- Uncategories), Antiepileptics, Barbiturates (Acidic/Neutral Drugs-Underivatized), Fentanyls, Organochlorine Pesticides III, Phenoxy Acid Herbicides, Styrene Impurities, TMS derivatized Sugars, Triazine Herbicides I, VOCs in Packaging materials.
- 14% cyqnopropyl,86% methyl polysiloxane phase
- Mid-polar
Ordering information: These columns are suitable replacements for DB-1701, Rtx-1701,CP-Sil 19CB, VF-1701.
Specifications
- Catalog No:
- 10510-864
- 10499-840
- 10499-842
- 10499-844
- 10499-854
- 10499-810
- 10499-856
- 10499-812
- 10499-858
- 10499-814
- 10499-816
- 10499-848
- 10499-850
- 10499-852
- 10499-808
- 10499-818
- 10499-820
- 10499-832
- 10499-834
- 10499-836
- 10499-860
- 10499-838
- 10499-824
- 10499-826
- 10499-828
- 10499-830
- 10499-870
- 10499-874
- 10499-876
- 10499-862
- 10499-864
- 10499-866
- 10499-868
- Description:
- 10510-864
- 10499-840
- 10499-842
- 10499-844
- 10499-854
- 10499-810
- 10499-856
- 10499-812
- 10499-858
- 10499-814
- 10499-816
- 10499-848
- 10499-850
- 10499-852
- 10499-808
- 10499-818
- 10499-820
- 10499-832
- 10499-834
- 10499-836
- 10499-860
- 10499-838
- 10499-824
- 10499-826
- 10499-828
- 10499-830
- 10499-870
- 10499-874
- 10499-876
- 10499-862
- 10499-864
- 10499-866
- 10499-868
- Temperature limit:
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 260/280°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 260/280°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 280/300°C
- -20 to 260/280°C
- -20 to 260/280°C
- -20 to 260/280°C
- -20 to 260/280°C
- -20 to 260/280°C
- -20 to 260/280°C
- -20 to 260/280°C