About this item
Typical application: Alcohols II and III, Amines (low MW), Amines/ Alcohols/ Chlorides, Aromatics analysis: ASTM D16 analytes, Aromatics II, Blood alcohol, Bourbon, Branched alcohols, BTEX, Chlorinated Isooctane, Citronella java oil, Esters III, Ethylene oxide, FAME Standard I, FAMEs (Coca Butter), Fast styrene analysis, Flavor volatiles, Formaldehyde Underivatized, Fragrance allergens, Fragrance Reference Standard II, Free Fatty Acids, Free Organic Acids/C4-C5 Isomers, Impurities in Ethylbenzene, Impurities in mixed xylenes, Impurities in p-Xylene-ASTM D3798, Impurities in styrene, Industrial Solvent, Lavender Oil Spiked with Camphor, Monomers in latex paint by headspace, Mushroom aroma (Synthetic), Nitrosamines, Nutmeg oil, Organic acids, Phenols III, Polyunsaturated fatty acid methyl esters, Rapeseed oil, Rapid analysis of water in organic solvents, Solvents I, II, Spearmint oil, Strawberry syrup, Sulphur and Selenium in Garlic by Headspace, Tramanian lavander oil, Volatile organics, Volatile sulfurs.
- Bondable polyethylene glycol
- Highest polarity and highest temperature limit of PEG phase
- Equivalent to USP G16
Specifications
- Catalog No:
- 10499-442
- 10499-444
- 10499-440
- 10499-480
- 10499-458
- 10499-482
- 10499-486
- 10499-452
- 10499-454
- 10499-456
- 10499-496
- 10499-498
- 10499-488
- 10499-490
- 10499-492
- 10499-494
- 10510-892
- 10510-894
- 10499-436
- 10499-348
- 10499-460
- 10499-438
- 10499-350
- 10499-352
- 10499-462
- 10499-354
- 10499-508
- 10499-430
- 10499-432
- 10499-434
- 10499-474
- 10499-518
- 10499-476
- 10499-478
- 10499-510
- 10499-466
- 10499-468
- 10499-512
- 10499-470
- 10499-514
- 10499-472
- 10499-516
- Description:
- 10499-442
- 10499-444
- 10499-440
- 10499-480
- 10499-458
- 10499-482
- 10499-486
- 10499-452
- 10499-454
- 10499-456
- 10499-496
- 10499-498
- 10499-488
- 10499-490
- 10499-492
- 10499-494
- 10510-892
- 10510-894
- 10499-436
- 10499-348
- 10499-460
- 10499-438
- 10499-350
- 10499-352
- 10499-462
- 10499-354
- 10499-508
- 10499-430
- 10499-432
- 10499-434
- 10499-474
- 10499-518
- 10499-476
- 10499-478
- 10499-510
- 10499-466
- 10499-468
- 10499-512
- 10499-470
- 10499-514
- 10499-472
- 10499-516
- Temperature limit:
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 50 to 220/240°C
- 50 to 220/240°C
- 50 to 220/240°C
- 50 to 220/240°C
- 40 to 240/250°C
- 40 to 240/250°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 50 to 220/240°C
- 40 to 240/250°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 240/250°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 240/250°C
- 40 to 260/280°C
- 40 to 260/280°C
- 40 to 240/250°C
- 40 to 260/280°C
- 50 to 220/240°C
- 40 to 260/280°C
- 50 to 220/240°C