Water ASTM Type II, deionized distilled, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: Hydrogen oxide
Formula:H₂O
MW:18.02 g/mol
Boiling Pt:100 °C (lit.)
Melting Pt:0 °C (lit.)
Density:1.000 g/ml at 3.98 °C (lit.)
MDL Number:MFCD00011332
CAS Number:7732-18-5
:Water
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 100 °C (lit.)
:Melting Pt:
:Melting Pt: 0 °C (lit.)
:Flash Pt:
:Flash Pt:
:M.W. 18.02 g/mol
:0 °C
:100 °C
Storage Temperature:Ambient
:12,10175
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Chemical formula:H₂O
- Molar Mass:18.02
- Storage:Room Temperature
- Chloride (Cl):5ppb max
- Conductivity at 25°C:1.0µS/cm max
- Expiration date:18 months from mfg date
- Form:Clear liquid
- Heavy metals (as Pb):0.01ppm max
- Iron (Fe):0.3ppm max
- Microbial limits:10 CFU/mL
- Resistivity at 25°C:1.0MOhm·cm min
- Silica (SiO2):3ppb max
- Sodium (Na):5ppb max
- Substances reducing permanganate:To pass test
- Total organic carbon:50µg/L max
- Total solids:20ppm max
- Supelco®, Water ASTM Type II, deionized distilled:
Specification Test Results
Customers Who Bought This Also Bought