2,3-Dichloro-5,6-dicyano-p-benzoquinone, Sigma-Aldrich®
Catalog #: CA8.02940.0025
Supplier: MilliporeSigma
CAS Number: 84-58-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:25 g
- Cat. no.:CA8.02940.0025
- Supplier No.:8.02940.0025
Specifications
Specification Test Results
- Hill Formula:C₈Cl₂N₂O₂
- Chemical formula:2,3-(Cl)₂-1-(=O)C₆-4-(=O)-5,6-(CN)₂
- Molar Mass:227 g/mol
- Melting Point:209 - 214 °C
- Solubility:(20 °C) (decomposition)
- LD 50 oral:LD50 Rat 86 mg/kg
- Storage class:6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
- WGK:WGK 3 highly water endangering
- R Phrase:R 25-29
- S Phrase:S 22-24/25-37-45
Specification Test Results
About this item
Synonyms: DDQ,2,3-Dichloro-5,6-dicyano-1,4-benzoquinone,2,3-Dichloro-5,6-dicyanoquinone,4,5-Dichloro-3,6-dioxocyclohexa-1,4-diene-1
ADR:6.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:M-Clarity™ quality level = MQ200
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 209 to 214 °C
:Flash Pt:
:Flash Pt:
:M.W. 227.01 g/mol
:396
UN:3439
:6.1
:III
:GHS06
:MQ200
Formula:C₈Cl₂N₂O₂
MW:227.01 g/mol
MDL Number:MFCD00001593
CAS Number:84-58-2
Customers Who Bought This Also Bought