To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Proteasome (prosome, macropain) 26S Subunit, non-ATPase 4
- Clonality:Monoclonal
- Clone:3G1A1
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:PSMD4
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:AF|ASF|S5A|Rpn10|pUB-R5|26S proteasome regulatory subunit S5A|Multiubiquitin chain binding protein|MCB1|26S proteasome non-ATPase regulatory subunit 4|AF-1|Antisecretory factor-1
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Storage temperature:Store at –20 °C
- Immunogen:PSMD4
- Purification:Protein G purification
- Cat. no.:10155-932
- Supplier No.:66179-1-IG
Specifications
About this item
The PSMD4 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human PSMD4. This antibody recognizes human antigen. The PSMD4 antibody has been validated for the following applications: ELISA, WB analysis.
WB 1 : 600
Type: Primary
Antigen: PSMD4
Clonality: Monoclonal
Clone: 3G1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human