To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Mucin 16, Cell Surface Associated
- Clonality:Monoclonal
- Clone:2B11B10
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Antigen symbol:MUC16
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Antigen synonyms:FLJ14303|CA125
- Storage buffer:PBS with 0.0.2% sodium azide and 50% glycerol pH 7.3.
- Storage temperature:Store at –20 °C
- Immunogen:MUC16,CA125
- Purification:Protein G purification
- Cat. no.:10155-872
- Supplier No.:60261-1-IG
Specifications
About this item
The MUC16,CA125 antibody from Proteintech is a mouse monoclonal antibody to a peptide of human MUC16,CA125. This antibody recognizes human,mouse antigen. The MUC16,CA125 antibody has been validated for the following applications: ELISA, IHC analysis.
WB 1 : 600
Type: Primary
Antigen: MUC16
Clonality: Monoclonal
Clone: 2B11B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse