To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:C1q tumor necrosis factor-related protein 3
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:C1QTNF3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Antigen synonyms:Collagenous repeat-containing sequence 26 kDa protein|CORS|CTRP3|Complement C1q tumor necrosis factor-related protein 3|C1ATNF3|CORS26|CORCS|CORS-26|UNQ753/PRO1484
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage temperature:Store at –20 °C
- Immunogen:C1QTNF3
- Purification:Antigen affinity purification
- Cat. no.:10154-462
- Supplier No.:11414-1-AP
Specifications
About this item
The C1QTNF3 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human C1QTNF3. This antibody recognizes human,mouse,rat antigen. The C1QTNF3 antibody has been validated for the following applications: ELISA, IHC analysis.
WB 1 : 1000 ; IHC 1 : 100
Type: Primary
Antigen: C1QTNF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat