L-Lysine, anhydrous 98.5-102.0%, Bioreagent
Supplier: VWR
Certificates
About this item
Synonyms: (S)-2,6-Diaminohexanoic acid,L(+)-Lysine,(S)-(+)-2,6-Diaminohexanoic acid,(S)-(+)-Lysine,(S)-(+)-2,6-Diaminocaproic acid
Formula:C₆H₁₄N₂O₂
MW:146.19 g/mol
MDL Number:MFCD00151035
CAS Number:56-87-1
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,5-102,0
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 196 °C
:Flash Pt:
:Flash Pt:
:M.W. 146.19 g/mol
EINECS:200-294-2
:196
:290
:12,05667
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Appearance:free flowing, white to off-white crystalline powder
- Assay (on dried basis, by titration):98.5% to 102.0%
- Identification (by IR):conforms to structure
- Residue On Ignition:NMT 0.1%
- Solubility (10% solution in H2O):clear, complete, colorless to yellow
- Specific Rotation (C=4g/100 mL H2O, calculated as free acid):+25.5º to +26.7º
- Water (by Karl Fischer):NMT 2.0%
- Cytotoxicity:passes test
- Bioburden:NMT 100 CFU/g
- Heavy Metals (as Pb):NMT 15 ppm
- Iron (Fe):NMT 30 ppm
- Sulfate (SO4):NMT 300 ppm
- DNase:none detected
- Endotoxin (1% solution in H2O):NMT 0.1 EU/mg
- RNase:none detected
- Protease:none detected
- L-Lysine, anhydrous 98.5-102.0%, Bioreagent:
Specification Test Results
Related Information
Certificates
Search by Lot NumberProduction Insight Solutions for Controlled Environments and Manufacturing Processes Production insight Solutions for Critical Environments and Manufacturing Processes Production insight v1 final1 VWR Bioprocessing Chemical and Excipients VWR Bioprocessing Chemicals and Excipients
Product Literature
More
spec_VWRB42307-100G spec_VWRB42307-1KG spec_VWRB42307.pdf
Product specification
Customers Who Bought This Also Bought