HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid, anhydrous ≥99.5%, Bioreagent
Supplier: VWR
Certificates
About this item
Synonyms: HEPES free acid,4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid,2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid,4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acid
Formula:C₈H₁₈N₂O₄S
MW:238.31 g/mol
Melting Pt:210…215 °C
Density:1.448 g/cm³ (20 °C)
MDL Number:MFCD00006158
CAS Number:7365-45-9
Merck Index:13,04672
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 210…215 °C
:Flash Pt:
:Flash Pt:
:M.W. 238.31 g/mol
:HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid
Storage Temperature:Ambient
EINECS:230-907-9
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Appearance:white crystalline powder
- Assay (By titration):NLT 99.5%
- Absorbance (1M solution in H2O) 260nm:NMT 0.10
- Absorbance (1M solution in H2O) 280nm:NMT 0.10
- Identification (by IR):conforms to structure
- pKa (@ 20ºC):7.45 to 7.65
- Residue On Ignition:NMT 0.3%
- Solubility (1M solution in H2O):clear, colorless, complete
- Water (KFT):NMT 0.5%
- Heavy Metals (as Pb):NMT 5 ppm
- Calcium (Ca):NMT 5 ppm
- Copper (Cu):NMT 5 ppm
- Iron (Fe):NMT 5 ppm
- Magnesium (Mg):NMT 5 ppm
- Manganese (Mn):NMT 5 ppm
- Bioburden:NMT 100 CFU/g
- Cell Culture Toxicity (at 12g/L):Passes test
- Endotoxin (0.2% solution in H2O):NMT 0.1 EU/mg
- DNase:none detected
- RNase:none detected
- Protease:none detected
- Additional Information:Retest Date (4 Yrs form Manufacture)
- HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid, anhydrous ≥99.5%, Bioreagent:
Specification Test Results
Related Information
Certificates
Search by Lot Number2015 Production insight V2 final Production Insight Solutions for Controlled Environments and Manufacturing Processes Production insight Solutions for Critical Environments and Manufacturing Processes Production insight v1 final1 VWR Bioprocessing Chemical and Excipients
Product Literature
More
spec_VWRB30487-1KG spec_VWRB30487-1KG spec_VWRB30487-25KG spec_VWRB30487-25KG spec_VWRB30487-5KG
Product specification
More
Customers Who Bought This Also Bought