Order Entry
ContactUsLinkComponent
Low-EDTA TE (1X), pH 8.0, Quality Biological
Low-EDTA TE (1X), pH 8.0, Quality Biological
Catalog #: 10128-588
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Low-EDTA TE 1X
Low-EDTA TE (1X), pH 8.0, Quality Biological
Catalog #: 10128-588
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • pH:
    8.0
  • Description:
    Low-EDTA TE (1X), pH 8.0
  • Size:
    4× 100 ml
  • Sterility:
    Sterile filtered
  • Shelf life:
    36 months
  • pH:
    8.0 ±0.1
  • Shipping temperature:
    Ambient
  • Supplier No.:
    351-324-721

Specifications

About this item

Low-EDTA 1X, pH 8.0 is molecular biology grade TE buffer used to store DNA and RNA. EDTA chelates Mg2+ and other divalent metals ions - (inhibits DNAse and RNAse to suppress DNA and RNA degradation).

  • With 0.1 mM EDTA
  • Filtered through a 0.1 µm membrane
  • DNase, RNase, and Protease tested

Tris is a buffering agent to keep the solution at a defined pH - The 'Cl' or 'HCl' indicates the pH was attained using aqueous hydrochloric acid (HCl), since Tris is naturally basic. Tris–EDTA (TE) is routinely used for suspending nucleic acid samples.

Low EDTA is recommended to re-suspend final purified DNA when DNA is used for downstream PCR.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.
Customers Who Bought This Also Bought