Protein Running Buffer 10X, Quality Biological
Catalog #: 10128-560
Supplier: QUALITY BIOLOGICAL, INC.
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Protein running buffer 10X
- Size:500 ml
- Sterility:Sterile filtered
- Test description:DNase, RNase and protease tested
- Shelf life:36 months
- Shipping temperature:Ambient
- Supplier No.:351-132-101
Specifications
About this item
Protein Running Buffer, 10X is used as the electrophoresis buffer during the stacking and resolve process of sodium dodecyl sulfate - polyacrylamide gel electrophoresis (SDS-PAGE), and is the running buffer for TRIS-glycine gel electrophoresis.
- Contains 1% SDS
- Filtered through a 0.22 µm membrane
- DNase, RNase, and Protease tested
Protein running Buffer, 10X is a Tris-Glycine buffer used for sodium dodecyl sulfate – polyacrylamide gel electrophoresis (SDS-PAGE) of proteins. It is used as both the anode and cathode buffer. Running Buffer, 10X is supplied as 500ml of 10X concentrate that can be diluted to a 1X solution.
This product is ideally suited for protein gel analysis in biochemical, molecular biological, cell biological, and immunological research studies.
Contents: Tris base, glycine, SDS and Molecular Biology Grade Water
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.
Customers Who Bought This Also Bought