Calcium chloride 2 M in aqueous solution, sterile filtered
Supplier: QUALITY BIOLOGICAL, INC.
Certificates
About this item
Synonyms: Chloro calcium
Formula:CaCl₂
MW:110.98 g/mol
MDL Number:MFCD00010903
CAS Number:10043-52-4
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 772 °C
:Flash Pt:
:Flash Pt:
:M.W. 110.98 g/mol
:Calcium chloride
EINECS:233-140-8
:772
:1600
:GHS07
Storage Temperature:Ambient
:14,01659
Specification Test Results
- in aqueous solution:
Specification Test Results
Customers Who Bought This Also Bought