Tris Glycine Transfer Buffer 10X (Transblotting Buffer 10X), Quality Biological
Catalog #: 10128-706
Supplier: QUALITY BIOLOGICAL, INC.
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Tris glycine transfer buffer 10X
- Size:1 L
- Sterility:Sterile filtered
- Test description:DNase, RNase and protease tested
- Shelf life:12 months
- Shipping temperature:Ambient
- Supplier No.:351-087-131
Specifications
About this item
TRIS Glycine Transfer Buffer, 10X (Transblot Buffer, 10X).
Tris Glycine Transfer Buffer, at a 1X concentration with the addition of 20% methanol, is used for electrophoretic transfer of proteins from denaturing polyacrylamide gel to a solid membrane – i.e. Western blotting or Tris-glycine gel electrophoresis.
Tris Glycine Transfer Buffer (10X) is prepared from molecular biology grade Tris base [tris(hydroxymethyl)- aminomethane] and glycine using Quality Biological’s Molecular Biology Grade (MBG) Water. The final product is sterile filtered using a 0.2 µm filter.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.
Customers Who Bought This Also Bought