Potassium chloride 1 M in aqueous solution, sterile filtered
Catalog #: 10128-494
Supplier: QUALITY BIOLOGICAL, INC.
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number: 7447-40-7
Specifications
- Size:500 ml
- Cat. no.:10128-494
- Supplier No.:351-044-101
Specifications
Specification Test Results
- Appearance:clear colorless liquid pass
- Dnase, Rnase and Protease:not detectable
- in aqueous solution:
Specification Test Results
About this item
Synonyms: Potassiumchloride
Density:1.13 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 773 °C
:Flash Pt:
:Flash Pt:
:M.W. 74.55 g/mol
EINECS:231-211-8
:773
:1420
Storage Temperature:Ambient
:12,07783
:14,07621
Formula:KCl
MW:74.55 g/mol
MDL Number:MFCD00011360
CAS Number:7447-40-7
Customers Who Bought This Also Bought