Sodium chloride 5 M in aqueous solution, sterile filtered
Supplier: QUALITY BIOLOGICAL, INC.
Certificates
About this item
Synonyms: Saline,Sea salt
Formula:NaCl
MW:58.44 g/mol
MDL Number:MFCD00003477
CAS Number:7647-14-5
:Sodium chloride
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 801 °C
:Flash Pt:
:Flash Pt:
:M.W. 58.44 g/mol
EINECS:231-598-3
:801
:1413
Storage Temperature:Ambient
:12,08742
:14,08599
Specification Test Results
- in aqueous solution:
Specification Test Results
Customers Who Bought This Also Bought