Magnesium chloride 1 M in aqueous solution, sterile filtered
Supplier: QUALITY BIOLOGICAL, INC.
Certificates
About this item
Formula:MgCl₂
MW:95.21 g/mol
MDL Number:MFCD00011106
CAS Number:7786-30-3
Merck Index:13,05684
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 715 °C
:Flash Pt:
:Flash Pt:
:M.W. 95.21 g/mol
:Magnesium chloride
EINECS:232-094-6
:715
:1412
Storage Temperature:Ambient
Specification Test Results
- Appearance:clear colorless liquid pass
- Dnase, Rnase and Protease:not detectable
- in aqueous solution:
Specification Test Results
Customers Who Bought This Also Bought