To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:YPAD Agar with Adenine Sulfate
- Size:40 mL
- Storage conditions:2°C to 8°C
- Sterility:Autoclaved
- Shipping conditions:Ambient Temperature
- Shelf Life:6 months
- Volume:100× 40 ml
- Supplier No.:342-310-219
Specifications
About this item
YPAD with Adenine Sulfate Agar is used to inhibit reversion of the ade1 and ade2 mutations.YPAD with Adenine Sulfate Agar inhibits red coloration in ade strains.
YPAD with Adenine Sulfate Agar is useful for detecting carcinogens with this yeast assay and preparing yeast stock for long-term storage.
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99