Order Entry
ContactUsLinkComponent
YPD/YEPD Plates, Quality Biological
YPD/YEPD Plates, Quality Biological
Catalog #: 10128-392
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
YPD/YEPD Plates CS100X25ml
YPD/YEPD Plates, Quality Biological
Catalog #: 10128-392
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    YPD/YPED Plates
  • Size:
    25 mL
  • Storage conditions:
    2°C to 8°C
  • Sterility:
    Autoclaved
  • Shipping conditions:
    Ambient Temperature
  • Shelf Life:
    6 months
  • Supplier No.:
    342-179-231

Specifications

About this item

YPD/YPED Plates are used for the growth of Saccharomyces cerevisiae (yeast).

A solid medium for the propagation of yeast.

UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only
Customers Who Bought This Also Bought