HyFlex® 11-900 Light Mechanical Protection Oil-Repellent Gloves, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
Providing good dexterity and tactility with light mechanical protection, these oil-repellent industrial work gloves are designed specifically for use in moderately oily manufacturing and maintenance environments.
- Dry-only grip
- Excellent abrasion resistance
- Secure, comfortable handling
- Superior tactile sensitivity with light mechanical protection
Gloves provide protection combined with a dry-only grip and abrasion resistance when workers need secure handling with comfort in these environments.
Use in the automotive and transportation, building and construction, machinery and equipment, maintenance, and metal fabrication industries. Uses include assembly of white goods; brown goods; light assembly tasks; and operating tools and power tools.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.
Specifications
- Catalog No:
- 80092-594
- 80092-588
- 80092-592
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- Nylon
- Nylon
- Nylon
- Finish:
- Palm Coated
- Palm Coated
- Palm Coated
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- 3131
- 3131
- 3131