Specifications
- Description:Dissolved oxygen/conductivity handheld instrument (must order the instrument, cable, and DO sensor separately)
- Supplier No.:6052030
Specifications
About this item
The Pro2030 provides everything you need in a handheld dissolved oxygen meter that automatically compensates for changes in salinity values.
- Rugged, simple and reliable
- Stores 50 data sets
- Graphic, backlit display and glow in the dark keypad: View display with polarized sunglasses on
- Quick Cal allows DO calibrations within seconds with the push of a button
- User replaceable DO sensors and cables (Choose field DO sensors (Polarographic or Galvanic))
- Built-in barometer and conductivity cell
- Measure DO, salinity, conductivity, specific conductance, TDS, barometric pressure and temperature
Ideal replacement for the discontinued YSI Model 85.
User-replaceable sensors and cables and convenient Quick Cal feature makes it extremely user friendly. Mil-spec (military spec) connectors, IP-67 waterproof and rubber over molded case, and 1-meter drop tests ensure the instrument remains in your hands to provide years of sampling even in the harshest field conditions. The feature set of the Pro2030 results in a low total cost of ownership over the life of the product and proves that it is smarter, tougher, and more versatile.
Fast response times with the Polarographic or Galvanic DO sensor allows you to complete your sampling routine quickly. The long life of the Polarographic sensor saves time and money and reduces your overall cost. The Galvanic sensor enables you to start sampling immediately without a warm-up period. Either sensor you choose for the field will give you accurate, quality data.
The super-stable 4-electrode conductivity cell is built into the cable and will provide years of accurate data. The cells are designed for rugged field performance and are easily cleaned and require minimal maintenance.
Available with 1, 4, 10, 20, or 30-meter cables (4-meter and longer cables include cable management kit). Fastest DO response time on the market with 95% response in 8 seconds with standard membrane. Multiple languages include English, Spanish, French and German.
3-year instrument; 2-year cable warranty.