BP624 GC Capillary Column, SGE
Supplier: Trajan Scientific and Medical
Certificates
About this item
More than five decades of innovative phase technologies and unique fused silica production capabilities, together provide end-to-end separation solutions for all applications.
- US EPA 624 optimized column
- Designed for volatiles analysis
- Able to be solvent rinsed
- Bonded and cross-linked
These columns can be used with EPA method 624, USP G43 methods, SW-846 and 8240/8260 methods. They are also for use with drinking water volatiles, chlorinated hydrocarbons and solvents.
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: These cyanopropylphenyl polysiloxane columns are suitable replacements for DB-624, OV-624, AT-624, HP-VOC, CP-Select624CB, 007-624, Rtx-Volatiles, VOCOL and ZB-624.
Specifications
- Catalog No:
- 60361-642
- 10799-630
- 14233-476
- 14233-478
- 14233-474
- 60361-634
- 60361-638
- 14233-480
- 60361-632
- Description:
- GC Capillary Column (Cyanopropylphenyl Polysiloxane)
- GC Capillary Column (Cyanopropylphenyl Polysiloxane)
- GC Capillary Column (Cyanopropylphenyl Polysiloxane)
- GC Capillary Column (Cyanopropylphenyl Polysiloxane)
- GC Capillary Column (Cyanopropylphenyl Polysiloxane)
- GC Capillary Column (Cyanopropylphenyl Polysiloxane)
- GC Capillary Column (Cyanopropylphenyl Polysiloxane)
- GC Capillary Column (Cyanopropylphenyl Polysiloxane)
- GC Capillary Column (Cyanopropylphenyl Polysiloxane)
- Temperature limit:
- 0 to 230/240°C
- 0 to 230/240°C
- 0 to 230/240°C
- 0 to 230/240°C
- 0 to 230/240°C
- 0 to 230/240°C
- 0 to 230/240°C
- 0 to 230/240°C
- 0 to 230/240°C