BPX35 GC Capillary Columns, SGE
Supplier: Trajan Scientific and Medical
Certificates
About this item
More than five decades of innovative phase technologies and unique fused silica production capabilities, together provide end-to-end separation solutions for all applications.
- Mid polarity column
- Ideal for confirmational analysis
- Inert and very low bleed
- Equivalent to USP phase G42
- High temperature
- Pharmaceutical specialist
- Bonded and cross-linked and able to be solvent rinsed
These columns can be used with environmental analyses, pesticides/herbicides, drugs of abuse, pharmaceuticals, polynuclear aromatic hydrocarbons and GC/MS applications.
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: These 35% phenyl polysilphenylene-siloxane columns are suitable replacements for DB-35, DB-35ms, Rtx-35, HP-35, HP-35MS, SPB-35 and MDN-35.
Specifications
- Catalog No:
- 60361-716
- 60361-670
- 60361-682
- 60361-714
- 60361-704
- 14226-020
- 14226-016
- Description:
- GC Capillary Column (35% Phenyl Polysilphenylene-siloxane)
- GC Capillary Column (35% Phenyl Polysilphenylene-siloxane)
- GC Capillary Column (35% Phenyl Polysilphenylene-siloxane)
- GC Capillary Column (35% Phenyl Polysilphenylene-siloxane)
- GC Capillary Column (35% Phenyl Polysilphenylene-siloxane)
- GC Capillary Column (35% Phenyl Polysilphenylene-siloxane)
- GC Capillary Column (35% Phenyl Polysilphenylene-siloxane)
- Temperature limit:
- 10 to 330/360°C
- 10 to 330/360°C
- 10 to 330/360°C
- 10 to 330/360°C
- 10 to 330/360°C
- 10 to 330/360°C
- 10 to 330/360°C