Specifications
- Description:Propylene glycol, 3.8 l (1 gal.)
- Temperature Range:–20...+100 °C (–4...+212 °F), when mixed 50/50 with distilled water
- Supplier No.:060320
Specifications
About this item
Propylene glycol is a safer alternative to ethylene glycol.
For use with VWR® Refrigerated Circulating Baths, VWR® Benchtop Chillers and VWR® Recirculating Chillers.
Lowers freezing point of water to allow circulation at below freezing temperatures. Mix with distilled water for broadest temperature range.