Specifications
- Description:Ethylene glycol, 3.8 l (1 gal.)
- Temperature Range:–25...+100 °C (–13...+212 °F), when mixed 50/50 with distilled water
- Supplier No.:060340
Specifications
About this item
A common circulator anti-freeze fluid.
For use with VWR® Refrigerated Circulating Baths, VWR® Benchtop Chillers and VWR® Recirculating Chillers.
Lowers the freezing point of water to allow circulation at below freezing temperatures. High toxicity. Recommend mixing with distilled water for broadest temperature range.