Cable Attachments, Brady Worldwide®
Supplier: Brady Worldwide
Certificates
About this item
Cable attachments are designed to lock out gate valves
Cable attachments are available in two formats. Choose between a 1/8" nonconductive nylon cable or a rustproof sheathed metal cable.
Customers Who Bought This Also Bought