Trajan, Phenyl Deactivated Fused Silica Capillary Tubing , Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
Certificates
About this item
Phenyl Deactivated Fused Silica Capillary tubing is available in a range of sizes.
- Chemically inert and thermally stable up to 380 °C (716 °F)
- Suitable for organic and aqueous solvents
Ideal for use in applications involving capillary GC guard columns and transfer lines.
Certifications: RoHS compliant per Directive 2011/65/EU.
Customers Who Bought This Also Bought