Order Entry
Canada
Orders LinkContactUsLinkComponent
Hematopoietic Progenitor Cell Medium, PromoCell

Hematopoietic Progenitor Cell Medium, PromoCell

Hematopoietic Progenitor Expansion Medium DXF, 500 ml
Catalog # 10172-388
Supplier:   Promocell
Supplier #:  C-28021
Hematopoietic Progenitor Cell Medium, PromoCell
Catalog # 10172-388
Supplier:  Promocell
CAS Number:  
Hematopoietic Progenitor Cell Medium, PromoCell
Hematopoietic Progenitor Cell Medium, PromoCell

Hematopoietic Progenitor Expansion Medium DXF, 500 ml

CAS Number:  
Price
$614.78
Log in for availability
Each (500ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The PromoCell Hematopoietic Progenitor Cell (HPC) Expansion Medium DXF has been designed for the maximum expansion of primitive HPCs in a fully defined, xeno-free culture environment

The PromoCell Hematopoietic Progenitor Medium has been developed for the expansion of hematopoietic progenitors by means of differentiation into more committed lineages of blood cells. Hence, this progenitor medium is suitable for CFU-based assays, e.g. long-term culture initiating cell (LTC-IC) assays.

Recommended for Human Mononuclear Cells (hMNC), Human CD34+ Progenitor Cells (hCD34+-CB), Human CD133+ Progenitor Cells (hCD133+-CB), Human Hematopoietic Progenitor Cells (HPCs)

Due to the utilization of exclusively synthetic, recombinant or plant-sourced materials, this medium is chemically defined and free of all animal-derived components and substances of human origin with Human Serum Albumin as the only exception. Note: The medium must be supplemented with additional cytokines, e.g. Cytokine Mix E (see below). Individual cytokines can be found on the PromoKine website.

Ordering information: The Hematopoietic Progenitor Expansion Medium DXF consists of a bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete Medium. In order to achieve optimal performance, the complete Medium must be supplemented with additional growth factors by the user. PromoCell Hematopoietic Progenitor Media contain all the growth factors and supplements necessary for the efficient expansion or directed differentiation of blood-derived progenitors into hematopoietic lineages.

Specifications

Specifications

  • Description:
    Hematopoietic Progenitor Expansion Medium DXF
  • Size:
    500 ml

Specifications

Accessories

Product Information

  • Catalog #
    Size
    Availability
    Price
  • Cytokine Mix E for HPC-Expansion Medium DXF, 100 mL
    Cytokine Mix E for HPC-Expansion Medium DXF, 100 mL
    10172-530
    100 ml
    Log in for availability
    Each - $1,135.69

Product Information

  • Catalog #
    Size
    Availability
    Price
  • Cytokine Mix E for HPC-Expansion Medium DXF, 500 mL
    Cytokine Mix E for HPC-Expansion Medium DXF, 500 mL
    10172-532
    500 ml
    Log in for availability
    Each (500ml) - $4,127.41

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.