Specifications
- Description:Osteoblast Mineralization Medium (Ready-to-use)
- Size:100 ml
- Cat. no.:10172-344
- Supplier No.:C-27020
Specifications
About this item
PromoCell Osteoblast Mineralization Medium was developed for the efficient induction of strong mineralization in human osteoblast cultures
Extensive extracellular calcium-deposits can be observed after 21 days, following an easy one-step protocol.