OTAL Hand Pumps, Bürkle
Supplier: BURKLE INC
Certificates
About this item
Delivery tubes made of PP are resistant against weak acids, caustic solutions and detergents.
Delivery tubes made of PVDF are especially for acids and caustic solutions; not for strongly oxidizing acids and caustic solutions.
Delivery tubes made of stainless steel 304 are for solvents and liquids of VbF Hazard Class AIII (not easily flammable).
Battery pump for safe filling of car batteries with acid or distilled water.
- Compact, space saving pumps
- With universal bungs for container openings of 30-70 mm dia.
- The OTAL mini pump made of PP (model 5005-1000) has a universal bung for container openings of 20-36 mm (0.79-1.42") dia.
Specifications
- Catalog No:
- 10216-964
- 10216-966
- 10216-952
- 10216-954
- 10216-956
- 76501-448
- 10216-958
- 10216-960
- 10216-962
- Flow rate:
- 12 L/min. (0.4 cfm)
- 5 L/min. (0.1 cfm)
- 4 L/min. (0.14 cfm)
- 14 L/min. (0.49 cfm)
- 20 L/min. (0.7 cfm)
- 14 L/min (3.70 gpm)
- 26 L/min. (0.9 cfm)
- 20 L/min. (0.7 cfm)
- 26 L/min. (0.9 cfm)