Barbed Reduction Couplers, Eldon James
Supplier: Eldon James Corp.
Certificates
About this item
These couplers are made of Natural Kynar® material, which has a temperature range of -62°C to 107°C (-80 to 226°F), and a pressure of 260 psi at 21°C (70°F)
This material can be autoclaved to 140°C (285°F) and meets USP Class VI requirements. Generally, chemically resistant to most acids, salts, weak bases, halogenated solvents, alcohols, nuclear and UV radiation, oxidants, ozone and deionized water.
Certifications: RoHS Compliant.
Related Information
Certificates
Search by Lot Number89238-112_1225095 89238-112_1246732 89238-118_1232792 89238-118_1256839 89238-118_1274645
Conformity declaration
More
Customers Who Bought This Also Bought