Barbed Adaptors, Eldon James
Supplier: Avantor
Certificates
About this item
These adapters are made of Natural Kynar® material.
- Chemically resistant
- Autoclavable
- Meets USP Class VI requirements
Natural Kynar® material has a temperature range of -62°C to 107°C (-80 to 226°F), and a pressure of 260 psi at 21°C (70°F). This material can be autoclaved to 140°C (285°F) and meets USP Class VI requirements. Generally, chemically resistant to most acids, salts, weak bases, halogenated solvents, alcohols, nuclear and UV radiation, oxidants, ozone and deionized water.
Certifications: RoHS Compliant. USP Class VI.
Related Information
Certificates
Search by Lot Number94025-248_1216052 94025-248_1216052 94025-250_1225089 94025-250_1225089 94025-252_1227327
Conformity declaration
More
Customers Who Bought This Also Bought