Distillation Glassware Set with Wheaton Connection®, Semi-Micro Kit™, Wheaton
Catalog #: 20762-096
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Description:Thermometer adapter, 20-400 screw threads
- Cat. no.:20762-096
- Supplier No.:165996
Specifications
About this item
Adapter has an “O” ring that holds a thermometer or gas inlet tube securely.
A grease-free WHEATON Connection® on one end provides an inert connection with external-threaded glass components. Adapter can also be used with gas inlet tubing and will accommodate any tube with an OD of 5 to 7 mm.
Customers Who Bought This Also Bought