Diazoxide 98%
Catalog #: AAJ66010-MC
Supplier: Thermo Scientific Chemicals
CAS Number: 364-98-7
Specifications
- Size:100 mg
- Cat. no.:AAJ66010-MC
- Supplier No.:J66010-MC
Specifications
Specification Test Results
- Assay:98%
Specification Test Results
About this item
Synonyms: 3-Methyl-7-chloro-1,2,4-benzothiadiazine 1,1-dioxide,7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1
:Diazoxide
Melting Pt:330…331 °C
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98%
:97,5
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 330…331 °C
:Flash Pt:
:Flash Pt:
:M.W. 230.666 g/mol
MW:230.666 g/mol
MDL Number:MFCD00078578
EINECS:206-668-1
:GHS07
Formula:C₈H₇ClN₂O₂S
Storage Temperature:Ambient
CAS Number:364-98-7
Customers Who Bought This Also Bought