Laerdal® Infant I.O. Training Leg
Catalog #: 470120-404
Supplier: American 3B Scientific
Specifications
- Description:Infant IO Training Leg
- Cat. no.:470120-404
- Supplier No.:W19565
Specifications
About this item
Use Alone, Or As Part Of The Nursing Baby Sim
- Simulated tibia and anatomical landmarks at the tibial tuberosity and medial malleolus
- Fluid may be infused for realistic flashback
- Drain in heel connects to reservoir bag
Lifelike reproduction of infant lower leg allows students to master infant intraosseous infusion techniques. Simulated tibia and anatomical landmarks at the tibial tuberosity and medial malleolus. Fluid may be infused for realistic flashback. Drain in heel connects to reservoir bag.
Customers Who Bought This Also Bought